Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106743.1 | NP_001100213.1 | 3 - 4 | 1163 | 71 | |
XM_006240323.2 | XP_006240385.1 | 3 - 4 | 1637 | 71 | ||
XM_006240324.2 | XP_006240386.1 | 3 - 4 | 1633 | 71 | ||
XM_006240325.2 | XP_006240387.1 | 3 - 4 | 1578 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |