Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297557.1 | NP_001284486.1 | 1 - 2 | 223 | 88 | |
NM_139081.2 | NP_620781.1 | 1 - 2 | 223 | 88 | ||
GenBank mRNA | D10706.1 | - | 1 - 2 | 195 | 88 | |
FQ211375.1 | - | 1 - 2 | 223 | 88 | ||
FQ213504.1 | - | 1 - 2 | 223 | 88 | ||
FQ213794.1 | - | 1 - 2 | 223 | 88 | ||
FQ214019.1 | - | 1 - 2 | 218 | 88 | ||
FQ214051.1 | - | 1 - 2 | 218 | 88 | ||
FQ214373.1 | - | 1 - 2 | 218 | 88 | ||
FQ214458.1 | - | 1 - 2 | 218 | 88 | ||
FQ216817.1 | - | 1 - 2 | 223 | 88 | ||
FQ218486.1 | - | 1 - 2 | 218 | 88 | ||
FQ218990.1 | - | 1 - 2 | 218 | 88 | ||
FQ220491.1 | - | 1 - 2 | 218 | 88 | ||
FQ220580.1 | - | 1 - 2 | 223 | 88 | ||
FQ225116.1 | - | 1 - 2 | 218 | 88 | ||
FQ225909.1 | - | 1 - 2 | 218 | 88 | ||
FQ225949.1 | - | 1 - 2 | 221 | 88 | ||
FQ229954.1 | - | 1 - 2 | 218 | 88 | ||
FQ230452.1 | - | 1 - 2 | 218 | 88 | ||
FQ231088.1 | - | 1 - 2 | 218 | 88 | ||
FQ232667.1 | - | 1 - 2 | 218 | 88 | ||
FQ232997.1 | - | 1 - 2 | 218 | 88 | ||
FQ233723.1 | - | 1 - 2 | 218 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |