Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ss18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100900.1 | NP_001094370.1 | 7 - 8 | 933 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |