Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108613.1 | NP_001102083.1 | 10 - 11 | 1136 | 59 | |
XM_006235769.2 | XP_006235831.1 | 9 - 10 | 3040 | 59 | ||
XM_006235771.2 | XP_006235833.1 | 10 - 11 | 3005 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |