Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191857.1 | NP_001178786.1 | 16 - 17 | 1590 | 63 | |
XM_006235772.2 | XP_006235834.1 | 16 - 17 | 1646 | 63 | ||
XM_006235773.2 | XP_006235835.1 | 16 - 17 | 2055 | 63 | ||
XM_006235774.2 | XP_006235836.1 | 16 - 17 | 1646 | 63 | ||
XM_006235775.2 | XP_006235837.1 | 16 - 17 | 1646 | 63 | ||
XM_006235776.2 | XP_006235838.1 | 16 - 17 | 1646 | 63 | ||
XM_006235778.2 | XP_006235840.1 | 9 - 10 | 1185 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |