Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfa2t2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168542.1 | NP_001162014.1 | 10 - 11 | 1611 | 71 | |
XM_006235311.2 | XP_006235373.1 | 10 - 11 | 1592 | 71 | ||
XM_006235312.2 | XP_006235374.1 | 11 - 12 | 1623 | 71 | ||
XM_008762313.1 | XP_008760535.1 | 10 - 11 | 1699 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |