Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035249.1 | NP_001030326.1 | 3 - 4 | 494 | 64 | |
GenBank mRNA | BC088226.1 | - | 3 - 4 | 412 | 64 | |
BC105907.1 | - | 3 - 4 | 494 | 64 | ||
BC126098.1 | - | 3 - 4 | 470 | 64 | ||
BC158564.1 | - | 3 - 4 | 478 | 8374563 | 64 | |
FQ214115.1 | - | 3 - 4 | 489 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |