Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025745.1 | NP_001020916.1 | 7 - 8 | 850 | 57 | |
XM_006245245.2 | XP_006245307.1 | 8 - 9 | 756 | 57 | ||
XM_006245246.2 | XP_006245308.1 | 8 - 9 | 738 | 57 | ||
GenBank mRNA | BC099215.1 | - | 7 - 8 | 850 | 7384656 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |