Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276401.1 | NP_001263330.1 | 3 - 4 | 475 | 63 | |
XM_006245206.1 | XP_006245268.1 | 3 - 4 | 410 | 63 | ||
XM_008767231.1 | XP_008765453.1 | 3 - 4 | 410 | 63 | ||
GenBank mRNA | FQ223111.1 | - | 3 - 4 | 395 | 63 |
Gene Symbol: LOC102556337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245329.1 | XP_006245391.1 | 3 - 4 | 410 | 63 | |
XM_006245336.1 | XP_006245398.1 | 3 - 4 | 410 | 63 | ||
XM_008767322.1 | XP_008765544.1 | 3 - 4 | 410 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |