Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107237.1 | NP_001100707.1 | 4 - 5 | 538 | 62 | |
XM_006251491.2 | XP_006251553.1 | 5 - 6 | 1311 | 62 | ||
XM_006251492.2 | XP_006251554.1 | 5 - 6 | 5146 | 62 | ||
XM_006251493.2 | XP_006251555.1 | 5 - 6 | 849 | 62 | ||
XM_006251494.2 | XP_006251556.2 | 5 - 6 | 1136 | 62 | ||
XM_006251495.2 | XP_006251557.1 | 5 - 6 | 824 | 62 | ||
XM_006251496.2 | XP_006251558.1 | 5 - 6 | 623 | 62 | ||
XM_006251497.2 | XP_006251559.1 | 5 - 6 | 857 | 62 | ||
XM_008770312.1 | XP_008768534.1 | 5 - 6 | 1136 | 62 | ||
XM_008770314.1 | XP_008768536.1 | 5 - 6 | 1136 | 62 | ||
XM_008770315.1 | XP_008768537.1 | 4 - 5 | 878 | 62 | ||
XM_008770317.1 | XP_008768539.1 | 4 - 5 | 878 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |