Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106416.2 | NP_001099886.1 | 2 - 3 | 365 | 71 | |
GenBank mRNA | BC166459.1 | - | 2 - 3 | 365 | 9027296 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |