Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012791.2 | NP_036923.1 | 8 - 9 | 1218 | 75 | |
XM_006248031.2 | XP_006248093.1 | 8 - 9 | 4163 | 75 | ||
XM_008768562.1 | XP_008766784.1 | 7 - 8 | 3426 | 75 | ||
XM_008768563.1 | XP_008766785.1 | 9 - 10 | 4299 | 75 | ||
XM_008768564.1 | XP_008766786.1 | 8 - 9 | 1265 | 75 | ||
XM_008768565.1 | XP_008766787.1 | 8 - 9 | 1340 | 75 | ||
GenBank mRNA | X79769.1 | - | 8 - 9 | 1220 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |