Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053447.1 | NP_445899.1 | 11 - 12 | 1426 | 97 | |
GenBank mRNA | AF321130.1 | - | 3 - 4 | 284 | 97 | |
BC161939.1 | - | 11 - 12 | 1401 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |