Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100757.1 | NP_001094227.1 | 1 - 2 | 83 | 84 | |
GenBank mRNA | BC169045.1 | - | 1 - 2 | 83 | 9100787 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |