Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109614.1 | NP_001103084.1 | 6 - 7 | 715 | 105 | |
XM_006241011.1 | XP_006241073.1 | 6 - 7 | 762 | 105 | ||
GenBank mRNA | BC158787.1 | - | 6 - 7 | 673 | 7304442 | 105 |
FQ220392.1 | - | 6 - 7 | 645 | 105 | ||
FQ229401.1 | - | 6 - 7 | 762 | 105 | ||
FQ230290.1 | - | 6 - 7 | 650 | 105 | ||
FQ234550.1 | - | 6 - 7 | 695 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |