Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tchh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232820.2 | XP_006232882.1 | 1 | 87 | 130 | |
XM_008761266.1 | XP_008759488.1 | 1 | 87 | 130 | ||
XM_008761267.1 | XP_008759489.1 | 1 | 87 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon |