Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013989.1 | NP_001014011.1 | 1 - 2 | 352 | 93 | |
XM_006250733.2 | XP_006250795.1 | 1 - 2 | 346 | 93 | ||
GenBank mRNA | BC079225.1 | - | 1 - 2 | 352 | 7132309 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |