Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037645.1 | NP_001032734.1 | 2 - 3 | 210 | 95 | |
XM_006251883.2 | XP_006251945.1 | 2 - 3 | 280 | 95 | ||
GenBank mRNA | BC107443.1 | - | 2 - 3 | 210 | 7307807 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |