Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017080.2 | NP_058776.2 | 3 - 4 | 656 | 73 | |
XM_006250471.2 | XP_006250533.1 | 4 - 5 | 437 | 73 | ||
XM_006250473.2 | XP_006250535.1 | 4 - 5 | 472 | 73 | ||
GenBank mRNA | AF542063.1 | - | 2 - 3 | 256 | 73 | |
BC078865.1 | - | 3 - 4 | 477 | 7109256 | 73 | |
FQ209646.1 | - | 3 - 4 | 423 | 73 | ||
FQ209829.1 | - | 3 - 4 | 423 | 73 | ||
FQ210302.1 | - | 3 - 4 | 421 | 73 | ||
FQ210596.1 | - | 3 - 4 | 423 | 73 | ||
FQ211059.1 | - | 3 - 4 | 420 | 73 | ||
FQ211404.1 | - | 3 - 4 | 422 | 73 | ||
FQ212763.1 | - | 3 - 4 | 424 | 73 | ||
FQ214152.1 | - | 3 - 4 | 423 | 73 | ||
FQ214397.1 | - | 4 - 5 | 479 | 73 | ||
FQ218558.1 | - | 3 - 4 | 421 | 73 | ||
FQ218584.1 | - | 3 - 4 | 423 | 73 | ||
FQ219241.1 | - | 3 - 4 | 422 | 73 | ||
FQ219385.1 | - | 3 - 4 | 421 | 73 | ||
FQ219394.1 | - | 3 - 4 | 421 | 73 | ||
FQ219517.1 | - | 3 - 4 | 423 | 73 | ||
FQ219567.1 | - | 3 - 4 | 432 | 73 | ||
FQ219624.1 | - | 3 - 4 | 356 | 73 | ||
FQ220122.1 | - | 3 - 4 | 421 | 73 | ||
J05107.1 | - | 3 - 4 | 404 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |