Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108594.1 | NP_001102064.1 | 1 - 2 | 106 | 81 | |
XM_006235128.2 | XP_006235190.1 | 1 - 2 | 146 | 81 | ||
GenBank mRNA | BC159413.1 | - | 1 - 2 | 89 | 8367451 | 81 |
FQ214547.1 | - | 1 - 2 | 78 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |