Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dio1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021653.3 | NP_067685.5 | 1 - 2 | 350 | 61 | |
GenBank mRNA | BC083557.1 | - | 1 - 2 | 345 | 7110283 | 61 |
CK480517.1 | - | 1 - 2 | 350 | 61 | ||
X57999.1 | - | 1 - 2 | 349 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |