Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053358.1 | NP_445810.1 | 3 - 4 | 535 | 125 | |
XM_006238532.2 | XP_006238594.1 | 3 - 4 | 570 | 125 | ||
XM_006238533.2 | XP_006238595.1 | 3 - 4 | 569 | 125 | ||
XM_006238534.2 | XP_006238596.1 | 3 - 4 | 569 | 125 | ||
XM_006238535.2 | XP_006238597.1 | 3 - 4 | 567 | 125 | ||
XM_006238536.2 | XP_006238598.1 | 3 - 4 | 568 | 125 | ||
XM_006238537.2 | XP_006238599.1 | 3 - 4 | 566 | 125 | ||
XM_006238538.2 | XP_006238600.1 | 3 - 4 | 566 | 125 | ||
XM_008763902.1 | XP_008762124.1 | 3 - 4 | 2223 | 125 | ||
XM_008763903.1 | XP_008762125.1 | 4 - 5 | 1777 | 125 | ||
XM_008763904.1 | XP_008762126.1 | 3 - 4 | 2220 | 125 | ||
XM_008763905.1 | XP_008762127.1 | 3 - 4 | 2220 | 125 | ||
XM_008763906.1 | XP_008762128.1 | 3 - 4 | 2217 | 125 | ||
XM_008763907.1 | XP_008762129.1 | 3 - 4 | 2219 | 125 | ||
XM_008763908.1 | XP_008762130.1 | 3 - 4 | 2217 | 125 | ||
XM_008763909.1 | XP_008762131.1 | 3 - 4 | 2214 | 125 | ||
GenBank mRNA | AF121893.1 | - | 3 - 4 | 535 | 125 | |
AY724490.1 | - | 3 - 4 | 235 | 125 | ||
BC107666.1 | - | 3 - 4 | 519 | 7936459 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |