Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022239.2 | NP_071575.1 | 6 - 7 | 483 | 152 | |
XM_006250872.2 | XP_006250934.1 | 5 - 6 | 1273 | 122 | ||
GenBank mRNA | M94555.1 | - | 6 - 7 | 483 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |