Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppcdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108763.1 | NP_001102233.1 | 2 - 3 | 252 | 81 | |
XM_008766303.1 | XP_008764525.1 | 2 - 3 | 344 | 81 | ||
XM_008766304.1 | XP_008764526.1 | 3 - 4 | 1825 | 81 | ||
XM_008766305.1 | XP_008764527.1 | 3 - 4 | 379 | 81 | ||
XM_008766306.1 | XP_008764528.1 | 3 - 4 | 403 | 81 | ||
XM_008766307.1 | XP_008764529.1 | 4 - 5 | 1105 | 81 |
Gene Symbol: NEWGENE_1306267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750512.1 | XP_003750560.1 | 250 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |