Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004247.1 | NP_001004247.1 | 6 - 7 | 634 | 97 | |
XM_006243155.2 | XP_006243217.1 | 6 - 7 | 602 | 97 | ||
XM_006243156.2 | XP_006243218.1 | 6 - 7 | 654 | 97 | ||
GenBank mRNA | BC079221.1 | - | 6 - 7 | 634 | 7131980 | 97 |
FQ209768.1 | - | 5 - 6 | 591 | 97 | ||
FQ212102.1 | - | 6 - 7 | 859 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |