Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005908.1 | NP_001005908.1 | 2 - 3 | 268 | 120 | |
XM_006252786.2 | XP_006252848.1 | 2 - 3 | 522 | 120 | ||
GenBank mRNA | BC083778.1 | - | 2 - 3 | 268 | 7193683 | 120 |
FQ212260.1 | - | 2 - 3 | 468 | 120 | ||
FQ217853.1 | - | 2 - 3 | 287 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |