Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243197.2 | XP_006243259.1 | 9 - 10 | 1049 | 70 | |
XM_006243198.2 | XP_006243260.1 | 10 - 11 | 1410 | 70 | ||
XM_006243199.2 | XP_006243261.1 | 9 - 10 | 1458 | 70 | ||
XM_006243200.2 | XP_006243262.1 | 9 - 10 | 1798 | 70 | ||
XM_006243201.2 | XP_006243263.1 | 9 - 10 | 3259 | 70 | ||
XM_008766374.1 | XP_008764596.1 | 9 - 10 | 929 | 70 | ||
XM_217153.7 | XP_217153.5 | 9 - 10 | 848 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |