Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017286.2 | NP_058982.1 | 3 - 4 | 622 | 61 | |
GenBank mRNA | BC089100.1 | - | 3 - 4 | 647 | 7307104 | 61 |
J05156.1 | - | 3 - 4 | 642 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |