Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_217183.8 | XP_217183.6 | 3 - 4 | 427 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |