Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109528.1 | NP_001102998.1 | 7 - 8 | 1069 | 72 | |
XM_006242959.2 | XP_006243021.1 | 7 - 8 | 1033 | 72 | ||
XM_006242960.2 | XP_006243022.1 | 7 - 8 | 1032 | 72 | ||
XM_006242961.2 | XP_006243023.1 | 7 - 8 | 981 | 72 | ||
XM_008766176.1 | XP_008764398.1 | 6 - 7 | 1231 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |