Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106581.1 | NP_001100051.1 | 8 - 9 | 925 | 102 | |
XM_006257018.2 | XP_006257080.1 | 7 - 8 | 992 | 102 | ||
XM_006257019.2 | XP_006257081.1 | 7 - 8 | 1003 | 102 | ||
XM_006257020.2 | XP_006257082.1 | 7 - 8 | 994 | 102 | ||
GenBank mRNA | BC169078.1 | - | 7 - 8 | 939 | 5598223 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |