Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014224.2 | NP_001014246.1 | 4 - 5 | 866 | 77 | |
GenBank mRNA | AB294402.1 | - | 4 - 5 | 866 | 77 | |
BC079040.1 | - | 4 - 5 | 938 | 7114771 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |