Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107864.2 | NP_001101334.2 | 7 - 8 | 933 | 71 | |
XM_006236599.1 | XP_006236661.1 | 8 - 9 | 1016 | 71 | ||
XM_006236600.2 | XP_006236662.1 | 8 - 9 | 1082 | 71 | ||
XM_006236601.2 | XP_006236663.1 | 7 - 8 | 2172 | 71 | ||
XM_008762979.1 | XP_008761201.1 | 7 - 8 | 2947 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |