Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145846.2 | NP_001139318.1 | 5 - 6 | 511 | 128 | |
NM_172077.2 | NP_742074.2 | 5 - 6 | 613 | 128 | ||
GenBank mRNA | AW532201.1 | - | 1 - 2 | 128 | ||
CF249035.1 | - | 4 - 5 | 128 | |||
D23676.1 | - | 5 - 6 | 511 | 128 | ||
L10229.1 | - | 5 - 6 | 613 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |