Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147177.1 | NP_671706.1 | 389 | 142 | ||
XM_006236865.2 | XP_006236927.1 | 463 | 142 | |||
GenBank mRNA | AB001581.1 | - | 376 | 142 | ||
AB002406.1 | - | 389 | 142 | |||
BC072511.1 | - | 374 | 7104777 | 142 | ||
BC086531.1 | - | 407 | 7304521 | 142 |
Gene Symbol: RGD1564855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773046.1 | XP_008771268.1 | 1 | 434 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |