Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153733.2 | NP_714955.2 | 2 - 3 | 230 | 99 | |
GenBank mRNA | BC061735.1 | - | 2 - 3 | 230 | 5598648 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |