Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153733.2 | NP_714955.2 | 4 - 5 | 439 | 146 | |
GenBank mRNA | AY075037.1 | - | 4 - 5 | 455 | 146 | |
BC061735.1 | - | 4 - 5 | 439 | 5598648 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |