Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127557.2 | NP_001121029.1 | 2 - 3 | 354 | 76 | |
XM_006256340.2 | XP_006256402.1 | 3 - 4 | 410 | 76 | ||
XM_006256341.2 | XP_006256403.1 | 3 - 4 | 463 | 76 | ||
GenBank mRNA | BC162057.1 | - | 2 - 3 | 356 | 7114691 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |