Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aire | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106379.1 | NP_001099849.1 | 9 - 10 | 1143 | 68 | |
XM_006256247.2 | XP_006256309.1 | 9 - 10 | 1222 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |