Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109672.1 | NP_001103142.1 | 4 - 5 | 395 | 105 | |
XM_006240549.2 | XP_006240611.1 | 5 - 6 | 638 | 105 | ||
XM_006240551.2 | XP_006240613.1 | 3 - 4 | 640 | 105 | ||
GenBank mRNA | BC129121.1 | - | 4 - 5 | 395 | 7125896 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |