Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040128.1 | NP_001035217.1 | 5 - 6 | 830 | 60 | |
XM_006227834.1 | XP_006227896.1 | 6 - 7 | 992 | 60 | ||
XM_006227837.2 | XP_006227899.1 | 6 - 7 | 1509 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |