Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191557.1 | NP_001178486.1 | 2 - 3 | 295 | 76 | |
XM_006255291.2 | XP_006255353.1 | 3 - 4 | 1845 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |