Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191558.1 | NP_001178487.1 | 12 - 13 | 1192 | 92 | |
XM_008772512.1 | XP_008770734.1 | 13 - 14 | 1303 | 92 | ||
XM_008772513.1 | XP_008770735.1 | 12 - 13 | 1253 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |