Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mysm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238409.2 | XP_006238471.1 | 9 - 10 | 1781 | 124 | |
XM_008763911.1 | XP_008762133.1 | 9 - 10 | 1400 | 124 | ||
XM_008763912.1 | XP_008762134.1 | 9 - 10 | 1400 | 124 | ||
XM_008763913.1 | XP_008762135.1 | 9 - 10 | 1400 | 124 | ||
XM_008763914.1 | XP_008762136.1 | 8 - 9 | 1376 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |