Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182814.2 | NP_877966.1 | 4 - 5 | 510 | 126 | |
GenBank mRNA | AY223861.1 | - | 4 - 5 | 376 | 126 | |
BC079283.1 | - | 4 - 5 | 481 | 7134210 | 126 | |
CA340557.1 | - | 4 - 5 | 516 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |