Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam73b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106566.1 | NP_001100036.1 | 7 - 8 | 980 | 95 | |
XM_006233781.1 | XP_006233843.1 | 7 - 8 | 1029 | 95 | ||
XM_006233782.2 | XP_006233844.1 | 7 - 8 | 1045 | 95 | ||
GenBank mRNA | FQ212119.1 | - | 7 - 8 | 948 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |