Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108577.2 | NP_001102047.1 | 7 - 8 | 863 | 147 | |
GenBank mRNA | BC166861.1 | - | 7 - 8 | 863 | 9101723 | 147 |
FQ223957.1 | - | 8 - 9 | 1018 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |