Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168549.1 | NP_001162021.1 | 12 - 13 | 4166 | 97 | |
XM_006233672.2 | XP_006233734.1 | 12 - 13 | 4134 | 97 | ||
XM_006233673.2 | XP_006233735.1 | 11 - 12 | 4101 | 97 | ||
XM_006233674.2 | XP_006233736.1 | 11 - 12 | 4101 | 97 | ||
XM_006233675.2 | XP_006233737.1 | 12 - 13 | 4134 | 97 | ||
XM_006233677.2 | XP_006233739.1 | 7 - 8 | 3046 | 97 | ||
GenBank mRNA | FQ211685.1 | - | 1 - 2 | 825 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |