Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spaca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191982.1 | NP_001178911.1 | 5 - 6 | 621 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |