Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108213.1 | NP_001101683.1 | 10 - 11 | 1839 | 161 | |
XM_006244819.2 | XP_006244881.1 | 11 - 12 | 2571 | 161 | ||
XM_006244820.2 | XP_006244882.1 | 11 - 12 | 3191 | 161 | ||
XM_006244821.2 | XP_006244883.1 | 10 - 11 | 2370 | 161 | ||
XM_008767030.1 | XP_008765252.1 | 7 - 8 | 1378 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |